Jenway 740201 Sipper Pump
Representative image only.
Jenway Item # EW-83056-41
Optimize performance of your Jenway® spectrophotometer
  • Expand analysis capabilities or make testing easier
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithJenway 7410, 7415, and 7615 Spectrophotometers
  • DescriptionSipper Pump
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Expand analysis capabilities or make testing easier
  • 1 year warranty
More About this Item
Choose from a wide range of accessories including communication cables, replacement bulbs, and more for your Jenway® spectrophotomer.
For safe and accurate sample handling the sipper pump will circulate samples and can be programmed to deliver reproducible sample volumes. Choose from manual or timed mode to accommodate any application.