InnoCal NIST-Traceable Calibration; Stroboscope
Representative image only.
InnoCal Item # EW-17050-10
  • Calibration service for stroboscopes
PRODUCT OPTIONS
$179.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeMechanical
  • System Calibration (Instrument and Probe)Yes
  • Number Of Calibration Test Points10 points
  • Calibration Test PointsThroughout range of instrument
  • Calibration Test Range0 to 120,000 flashes per minute
  • DescriptionNIST-Traceable Calibration; Stroboscope
Key Features
  • Calibration service for stroboscopes
More About this Item
Calibration reports issued by InnoCal may not be reproduced, except in full, without written permission of InnoCal. The results stated in our reports relate only to the items tested or calibrated. Measurements reported on our reports are traceable to SI units via national standards maintained by NIST and are performed in compliance with MIL-STD-45662A, ANSI/NCSL Z540-1-1994, 10CFR50, Appendix B, ISO9002-94, and ISO/IEC 17025:2005
InnoCal's Metrology Laboratory is accredited to ISO/IEC 17025 by the American Association for Laboratory Accreditation (A2LA); certificate number 1746.01. This accreditation is your assurance that one of the foremost outside auditing agencies approves of our internal laboratory policies and procedures.
Please see part number 17050-00 for combination contact/non-contact tachometers.