Keeping humidity instruments calibrated helps you measure and regulate this variable more accurately
Testing range from 5 to 95% RH
Standard testing at three points: 30, 60, and 80% RH with one temperature point at ambient (22 to 25°C), with customized tests available upon request
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s), and test uncertainty ratios (TUR’s) at no additional cost
This NIST-traceable calibration allows you to confirm that the specific asset you purchase meets the manufacturer’s published specification
EPA, USDA, ISO, and GLP/GMPs standards require regular NIST-Traceable calibration of all test equipment being used in critical applications, FDA compliant
Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays
All measurements reported are traceable to SI units via national standards maintained by NIST and performed in compliance with ANSI Z540, ISO17025, and MIL-STD-45662A
Testing range from 5 to 90% RH at uncertainties as low as
(K=2) ±0.59% RH
Testing at three points: 30, 60, and 80% RH with one temperature point
at ambient (22 to 25C)
NIST-Traceable calibration for relative humidity and temperature recorders
Standard testing at three points: 30, 60, and 80% RH with one temperature point at ambient (22 to 25°C), with customized tests available upon request
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s), and test uncertainty ratios (TUR’s) at no additional cost
This NIST-traceable calibration allows you to confirm that the specific asset you purchase meets the manufacturer’s published specification
EPA, USDA, ISO, and GLP/GMPs standards require regular NIST-Traceable calibration of all test equipment being used in critical applications, FDA compliant
Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays
All measurements reported are traceable to SI units via national standards maintained by NIST and performed in compliance with ANSI Z540, ISO17025, and MIL-STD-45662A
Testing range from 5 to 90% RH at uncertainties as low as
(K=2) ±0.59% RH
Testing at three points: 30, 60, and 80% RH with one temperature point
at ambient (22 to 25C)
NIST-Traceable calibration for relative humidity and temperature recorders
More About this Item
From humidity data loggers to thermohygrometers to high-accuracy handheld instruments, InnoCal offers humidity calibrations for nearly any brand or style of meter from 0 to 95% relative humidity (RH). InnoCal services all major brands of hygrometers including Vaisala, Cole-Parmer, Extech, Oakton, Fluke, and many more! InnoCal is accredited to ISO/IEC 17025 by the American Association for Laboratory Accreditation (A2LA) under certificate number 1746.01. This accreditation is your assurance that the calibration is performed in a qualified, controlled laboratory.
Our highly-trained technicians provide NIST-traceable accredited humidity calibrations using Condensation Chilled Mirror Hygrometer technology in a dedicated environmental chamber.