InnoCal NIST-Traceable Calibration, Process Calibrator/Digital Multimeter
Representative image only.
InnoCal Item # EW-17101-78
NIST-traceable calibrations for multimeters, calibrators, oscilloscopes, and more!
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Calibration service for a process calibrator or digital multimeter
PRODUCT OPTIONS
$208.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Calibration TypeElectrical
  • System Calibration (Instrument and Probe)Yes
  • Calibration Test PointsCalibrated per manufacturer's procedure
  • DescriptionNIST-Traceable Calibration; Process Calibrator / Digital Multi-Meter
NIST Traceable Calibration
Key Features
  • Provides traceability to NIST
  • Lists the environmental conditions present at time of calibration
  • Certificate lists actual values, uncertainties, and tolerances
  • Regular calibration helps you meet ISO, EPA, GLPs/GMPs and other quality standards
  • Calibration service for a process calibrator or digital multimeter
More About this Item
All calibration certificates contain as-found and as-left data, estimated measurement uncertainties (emu’s) and test uncertainty ratios (TUR’s) at no additional cost!
NIST-traceable calibration allows you to confirm that the specific asset you purchase does in fact meet the manufacturer’s published specification. Receive your product calibrated and ready to use rather than sending it to a third-party or internal calibration lab and incurring additional delays. EPA, USDA, ISO and GLP/GMP standards require regular NIST-Traceable calibration of all test equipment being used in critical applications.
Calibrations on handheld process calibrators are performed using high-accuracy multi-function calibrators and standard-grade reference multimeters per manufacturer’s recommended procedures. Measurements can be taken up to 1,000 Volts, 20 Amps, 1100 Mega-Ohms, and 1.1 Giga-Hertz. Temperature source and measurements can be performed between -250 and 630OC on 8 types of RTDs and -250 to 2316OC on 11 types of thermocouples.