InnoCal NIST-Traceable Calibration; Calibration Weight/Mass, 1 to 25 kg; ASTM 1-3; OIML F1/F2
Representative image only.
InnoCal Item # EW-17108-18
Mass Value Report
  • Measurements are traceable to NIST
  • Certificate lists actual weight values, uncertainties, and tolerances
PRODUCT OPTIONS
$104.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionNIST Calibration Service for Masses. 1 to 25kg; ASTM 1-3; Oiml F1/F2
Key Features
  • Measurements are traceable to NIST
  • Certificate lists actual weight values, uncertainties, and tolerances
More About this Item
Mass calibrations are performed by an ISO 17025 accredited sub-contractor. InnoCal, our internal ISO 17025 accredited laboratory manages this sub-contracted service for our customers ensuring that the strictest quality requirements are being maintained. Calibration reports are issued directly to the customer after incoming inspection is performed by InnoCal's metrology staff. All measurements reported are traceable to SI units.