Inlet/Outlet filter, 10 micron filter, 31.8 CFM, 1/2" NPT(F)

Representative image only.
Mfr # PPSF.5-X35 – Item # EW-79700-70
Protect your pump and equipment from dust and particles
  • Use on either with vacuum down to 29" Hg or pressure up to 150 PSI
  • 10-Micron filter element is easily replaced
  • Shatterproof clear plastic housing

PRODUCT OPTIONS

$48.25USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Connections1/2" NPT(F)
  • Length (in)5.125
  • Width (in)3.5
  • DescriptionInlet/Outlet filter, 10 micron filter, 31.8 CFM, 1/2" NPT(F)

Key Features

  • Use on either with vacuum down to 29" Hg or pressure up to 150 PSI
  • 10-Micron filter element is easily replaced
  • Shatterproof clear plastic housing

More About this Item

Wetted materials include a nylon head, Buna-N gasket, polyethylene (PE) element, and a polycarbonate bowl. Stainless steel elements are also available; see ordering table.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat