IKA RV 3 V Rotary Evaporator With Dry Ice Condenser, Uncoated Glassware, And Digital Temperature Control; 100 To 240 VAC

Representative image only.
IKA Mfr # 0010003494 – Item # EW-28711-32
Updated design features digital displays for speed and temperature with a larger 4 L bath
  • Integrated safety function lifts flask out of bath in case of power failure
  • Red warning light indicates unlocked position of vapor tube
  • Single-handed manual lift offers precise positioning of glassware
  • Low device voltage (24 V) ensures user safety
  • Adjustable immersion angle
  • Includes a dry ice condenser and uncoated glassware
  • 5 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Display TypeDigital
  • Condenser TypeCold-trap
  • Min Speed (rpm)20
  • Max Speed (rpm)300
  • Glass TypeUncoated
  • Lift MechanismManual
  • Bath TypeWater/Oil
  • Bath Capacity (L)4
  • Min Temperature (° C)20
  • Max Temperature (° C)90
  • Power (VAC)100 to 240
  • Power (watts)1400
  • Power (Hz)50/60
  • Width (in)17 3/8
  • Height (in)20 7/8
  • Depth (in)13
  • Width (cm)44
  • Height (cm)53
  • Depth (cm)33
  • DescriptionRotary Evaporator With Dry Ice Condenser, Uncoated Glassware, And Digital Temperature Control; 100 To 240 VAC
  • Warranty5 Years
5 Year Warranty

Includes

Heating bath, condenser set, evaporating and condensing flasks, Woulff bottle, and power cord.

Key Features

  • Integrated safety function lifts flask out of bath in case of power failure
  • Red warning light indicates unlocked position of vapor tube
  • Single-handed manual lift offers precise positioning of glassware
  • Low device voltage (24 V) ensures user safety
  • Adjustable immersion angle
  • Includes a dry ice condenser and uncoated glassware
  • 5 year warranty

More About this Item

These rotary evaporators are designed and engineered for ease of use, while maintaining the highest level of safety. Manual lift handling can be done with one hand for precise positioning of glassware and is suitable for left- or right-handed operators. A flask clamping mechanism with integrated push-off function allows for easy exchange of evaporation flasks. The transparent vertical condenser's small footprint makes efficient use of the 1500 cm² cooling surface.

The unique 4 L bath design provides fast heat up, allowing for quicker experiments. Non-exposed heating coils and integrated handles offer safe operation. Bath heats from 20 to 90°C, adding versatility by allowing use of water or oil. The DC motor provides rotation speed from 20 to 300 rpm. Each unit comes standard with a dual digital display for both bath temperature and rotational speed to provide more control and easier repetition of experiments.

The evaporators are available with or without safety-coated glassware to minimize breakage and increase safety. Safety coating resists most solvents and acids while protecting you against shattered glass and dangerous reagents due to broken glass.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat