ICI Quick Ship Envision Female Quick-Connect Fittings, Nitrogen, Brown
Representative image only.
ICI Quick Ship Mfr # CPQCFL-B-3M-K8 – Item # EW-78984-46
Quick connect fittings designed specifically for Envision workstation plumbing fixtures
  • Conveniently color labeled for easy coupling
  • Available for air, vacuum, and nitrogen services
  • Include all hardware necessary to attach to the service panel
  • 2 year warranty
PRODUCT OPTIONS
$251.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ColorBrown
  • Compatible With78984-40 & 78984-43
  • DescriptionEnvision Female Quick-Connect Fittings, Nitrogen, Brown
  • Warranty2 Years
Two year warrantyMade in the U.S.A.
Key Features
  • Conveniently color labeled for easy coupling
  • Available for air, vacuum, and nitrogen services
  • Include all hardware necessary to attach to the service panel
  • 2 year warranty
More About this Item
ICI Quick Ship female quick disconnect fittings are designed to mate with the Envision plumbing assemblies. Simply mount with the overhead service panel and connect it from the workstation to the building services. Fittings are designed to only couple with the same key, which is conveniently color coded in orange, yellow, and brown. All fittings include 3/8 in stems and hardware to attach to the overhead service panel.