Humboldt H-6220 Accuflame Bunsen Burner, TIRrill Burner, Natural Gas
Representative image only.
Humboldt Mfr # H-6220 – Item # EW-36130-10
  • 3 month warranty
PRODUCT OPTIONS
$77.50USD / Each
BUY MORE & SAVE
  • 1-9$77.50USD / Each
  • 10-49$69.75USD / Each
  • 50-99$65.88USD / Each
  • 100+$62.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Gas TypeNatural Gas
  • BTU Output (per hour)1200
  • Inlet Tube OD (in)0.4375
  • Height (in)6
  • DescriptionAccuflame Bunsen Burner, TIRrill Burner, Natural Gas
  • Warranty3 Months
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Straight Tube Gas Burners feature adjustable flames for very slow heating or quick boiling. Flame retainer prevents flame lifting or extinguishing. Tapered serrated gas inlet connections for easy tube
mounting.