Humboldt H-5810 Accuflame Bunsen Burner, Micro-Burner, Natural Gas
Representative image only.
Humboldt Mfr # H-5810 – Item # EW-36130-30
  • 3 month warranty
PRODUCT OPTIONS
$68.00USD / Each
BUY MORE & SAVE
  • 1-9$68.00USD / Each
  • 10-49$61.20USD / Each
  • 50-99$57.80USD / Each
  • 100+$54.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Gas TypeNatural Gas
  • BTU Output (per hour)1200
  • Inlet Tube OD (in)0.3125
  • Height (in)3 1/2
  • DescriptionAccuflame Bunsen Burner, Micro-Burner, Natural Gas
  • Warranty3 Months
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Microburners include adjustable air supply and tapered serrated gas connection. Optional tripod stand 36310-50 supports a 1-L beaker.