Howard Leight MAXIMUM Disposable Earplugs with Cord
Representative image only.
Howard Leight Mfr # MAX-30 – Item # EW-86310-01
  • 1 year warranty
PRODUCT OPTIONS
$69.90USD / Box of 100
BUY MORE & SAVE
  • 1-9$69.90USD / Box of 100
  • 10+$62.91USD / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • NRR (dB)33
  • ComplianceANSI S3.19-1974; CSA class A(L)
  • CordedYes
  • DescriptionDisposable Earplugs with cord
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Maximum 33 dB attenuation is ensured when you use these pre-shaped foam ear plugs. The smooth, soil resistant outer skin prevents dirt from penetrating the surface prior to insertion. Individually wrapped by pairs in poly-bags.