Howard Leight MAXIMUM LITE Earplugs, Corded; 100 Pairs/Box
Representative image only.
Howard Leight Mfr # LPF-30 – Item # EW-86310-71
Low-pressure earplugs offer long-term comfort and protection
  • Non-irritating
  • Non-allergenic
  • Individually wrapped by pairs in poly bags
PRODUCT OPTIONS
$61.30USD / Box of 100
BUY MORE & SAVE
  • 1-9$61.30USD / Box of 100
  • 10+$55.17USD / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • NRR (dB)30
  • ComplianceANSI S3.19-1974; CSA class A(L)
  • CordedYes
  • DescriptionEarplugs, Corded; 100 Pairs/Box
Key Features
  • Non-irritating
  • Non-allergenic
  • Individually wrapped by pairs in poly bags
More About this Item
The self-adjusting foam recovers to fit vitually any size ear canal.