Honeywell T105310 Filter cartridge with P100 filter; Organic vapor/acid gas
Representative image only.
Honeywell Mfr # T105310 – Item # EW-81661-63
Efficient, dependable protection
  • For use with Survivair™ T-Series Respirators, acid gas, organic vapors, and P100
  • Easy to install and replace
  • Meets NIOSH Approved standards
SPECS & DESCRIPTION
Specifications & Description
  • RatingP100
  • NIOSH ApprovalCertain organic vapors, Chlorine, Hydrogen Chloride, Sulfur Dioxide, Chlorine Dioxide, Hydrogen Fluoride, Hydrogen Sulfide (escape only)
  • ComplianceNIOSH
  • ColorPink, Yellow
  • DescriptionFilter cartridge with P100 filter; Organic vapor/acid gas
Key Features
  • For use with Survivair™ T-Series Respirators, acid gas, organic vapors, and P100
  • Easy to install and replace
  • Meets NIOSH Approved standards
More About this Item
Honeywell T-Series respirator cartridges offer a 99.7% filter efficiency protecting against acid gases and organic vapors. It resists airborne particulates, chemicals, contamination, gas, and vapors and smoke. The lock-twist cartridges provide an audible click indicating a positive lock on the connector.