Hollow Fiber Filter, 0.2 um, Female/Male Luer; 12/Pk
Representative image only.
Item # EW-29510-50
Reduce the risk of contamination to continuous perfusion cultures
  • Made of polysulfone housing and end caps
  • Features polypropylene autovent to prevent air locking
  • Operate filter in any position
  • Nonsterile
  • Autoclavable
  • 1 year warranty
$943.00USD / Box of 12
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialMixed Cellulose Ester
  • Pore Size (micron)0.2
  • Housing MaterialPolysulfone
  • ConnectionsFemale luer lock by male luer lock
  • Effective Filtration Area (cm²)70
  • Surface Area (m²)0.007
  • Max Operating Temperature (° C)45
  • Max Operating Temperature (° F)113
  • Max Pressure (PSI)30
  • Max Pressure2
  • Min Throughput (Liters)2
  • Max Throughput (Liters)5
  • Min Throughput (mL)2000
  • Max Throughput (mL)5000
  • DescriptionHollow Fiber Filter, 0.2 um, Female/Male Luer; 12/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Made of polysulfone housing and end caps
  • Features polypropylene autovent to prevent air locking
  • Operate filter in any position
  • Nonsterile
  • Autoclavable
  • 1 year warranty
More About this Item
Ideal for bioprocessing, fermentation, and cell culture applications. Naturally hydrophilic cellulose hollow fiber membrane exhibits low extractables and high biocompatibility. Use two filters in series to form a sterile barrier when feeding media to the bioreactor. Easily change the first filter if it becomes plugged while the second continues to maintain culture sterility. Includes 1/8" hose adapters and clamps.