Hirschmann pipetus® Pipette Controller, 7.1 mL/sec; 100-240 VAC
Representative image only.
Hirschmann Mfr # 9907200 – Item # EW-22800-04
Pipette filler for all pipettes from 0.1 mL up tp 200 mL
  • Digital, color, display with battery level indicator
  • 1 year warranty
$495.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • AutoclavableYes
  • Number of Speeds5
  • Speed7.1mL/sec
  • Power (VAC)120 / 220
  • Battery1 NiMH (included)
  • DescriptionPipette Controller, 7.1 mL/sec; 100-240 VAC
  • Warranty1 year
1 Year Warranty
Includes
universal AC power adapter/charger with 3 plugs (EU, US, and UK), 1 rechargeable NiMH battery, safety valve, two filters, and charging stand for inductive charging.
Key Features
  • Digital, color, display with battery level indicator
  • 1 year warranty
More About this Item
Noise-free, lightweight, and powerful Pipetus® Pippetting Controller ensures precise results with sensitive valves for exact adjustments of the meniscus. The EX/Blow out key features both motor driven and gravity run-out of pipette contents. Five-stage speed control allows flexibility when working with changing volumes and viscosity.