Heavy-Duty HDPE Crock with Cover, 4 gal; 1/Pk

Representative image only.
Item # EW-06724-40

$135.00USD / Each

BUY MORE & SAVE

  • 1-5$135.00USD / Each
  • 6-29$121.50USD / Each
  • 30-119$114.75USD / Each
  • 120+$108.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (L)15
  • Capacity (Gal)4
  • MaterialHDPE
  • Top Outside Diameter (in)12
  • Top Outside Diameter (cm)30.5
  • Overall Height (in)13 1/4
  • Overall Height (cm)33.7
  • DescriptionHeavy-Duty HDPE Crock with Cover, 4 gal; 1/Pk

More About this Item

These sturdy, one-piece molded units are made from high-density polyethylene (HDPE) and replace conventional crocks. Ideal for use with round dipping baskets. All crocks include cover. NSF certified; USDA approved.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat