Hayward Vent Valve, Vacuum Breaker, 3/4" NPT (F), PVC
Representative image only.
Hayward Mfr # VB107X15 – Item # EW-06225-95
Eliminate problems when evacuating closed tanks or piping systems
  • Constructed of durable PVC with a Viton® seal
$214.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPVC
  • Connection TypeNPT(F)
  • Thread size NPT(F) (port 1) (in)3/4
  • DescriptionVent Valve, Vacuum Breaker, 3/4" NPT (F), PVC
Key Features
  • Constructed of durable PVC with a Viton® seal
More About this Item
Eliminates problems when evacuating closed tanks or piping systems. The one-way valve that is normally closed at positive pressure will allow drainage at a rate of 300 GPM. For higher drainage rates use additional vacuum breakers. The 3⁄4" NPT(F)socket adapts easily onto a 11⁄2" bulkhead fitting. The breaker is made of PVC with a Viton® seal.