Hayward TC10050STE True Union Ball Check Valve, 1/2" socket and NPT(F) connectors
Representative image only.
Hayward Mfr # TC10050STE – Item # EW-01341-00
Unique square-cut elastomer seat provides seal at very low back pressures
PRODUCT OPTIONS
$63.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.5
  • Outlet Port1/2" NPT(F) and socket
  • Connection TypeNPT(F) and Socket
  • Inlet Port1/2" NPT(F) and socket
  • Cv4.6
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC, EPDM
  • Working Pressure (PSI)150
  • DescriptionTrue Union Ball Check Valve, 1/2" socket and NPT(F) connectors
More About this Item
Quickly and easily install either vertically or horizontally. These valves are assembled with water soluble, silicone-free lubricant and constructed of a one-piece PVC body.