Hayward 155.522.554 PVC Needle Valve, 1/4" NPT(F), with PTFE and Viton
Representative image only.
Hayward Mfr # NVA1025T – Item # EW-03245-60
Don’t worry about leaks
  • Accurate flow control lets you adjust flow down to drops per minute
  • Seals are heavy-duty FPM O-ring
  • Natural PVDF valves are the ideal option for high-purity, high-temperature applications
PRODUCT OPTIONS
$92.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Port Size (NPT(F))0.25
  • Outlet Port1/4" NPT(F)
  • Connection Type1/4" NPT(F)
  • Inlet Port1/4" NPT(F)
  • Cv0.64
  • Housing MaterialPVC
  • Max Pressure (PSI)150
  • Max Pressure (bar)10.3
  • Max Temperature (° F)140
  • Max Temperature (° C)60
  • Wetted MaterialsPVC
  • DescriptionPVC Needle Valve, 1/4" NPT(F), with PTFE and Viton
Key Features
  • Accurate flow control lets you adjust flow down to drops per minute
  • Seals are heavy-duty FPM O-ring
  • Natural PVDF valves are the ideal option for high-purity, high-temperature applications
More About this Item
Precision needle valves are available in four materials: PVC, CPVC, PP and PVDF. All feature an integrated stem with PTFE seat; fine pitch stem threads ensure precise adjustment. Valve body also includes flanges for panel mounting. Connections are standard pipe thread.