Haws 7656WCSM Pull-Down Eyewash, Wall-Mount
Representative image only.
Haws Mfr # 7656WCSM – Item # EW-06767-51
Inverted flow of eyewash sweeps contaminants away from inner eye and nasal cavity
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Eyewash Flow Rate (GPM)3.7
  • Bowl MaterialStainless steel
  • MountWall Mount
  • Outlet Connections2\" NPT(F)
  • Inlet Connection1/2\" NPT(M)
  • ComplianceANSI Z358.1-2009
  • DescriptionPull-Down Eyewash, Wall-Mount
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Pull-down eyewash features a unique inverted flow to push contaminants away from the inner corner of the eye, consistent with first aid protocols. With other eyewashes, contaminants can travel through the inner corner of the eye to the nasal cavity resulting in further injury. Type 304 stainless steel cabinet is surface mounted for installation wherever you need it. Simply pull down the eywash to activate water flow.
Compliance: ANSI Z358.1-2009