Hardman Double Bubble® Epoxy Pouches, High Peel Strength, 3.5g; 10/Pk
Representative image only.
Hardman Mfr # 4007-BG10 – Item # EW-08778-08
Complete selection of epoxies and urethane for all your maintenance needs
  • Single-use adhesives are perfect for quick repair jobs
  • A flexible, tough and durable, vibration resistant adhesive
  • High peel and shear strengths
  • Bonds to polystyrene, ABS nylon, metal, wood, masonry and rubber
PRODUCT OPTIONS
$37.50USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$37.50USD / Pkg of 10
  • 10+$33.75USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Curing time (days)7
  • Curing time (hrs)96
  • Set time (mins)240
  • Set time (hrs)4
  • Min operating temperature (° F)-40
  • Max operating temperature (° F)250
  • Number of Pieces per Set10
  • DescriptionEpoxy Pouches, High Peel Strength, 3.5g; 10/Pk
Includes
10 pouches and mixing sticks.
Key Features
  • Single-use adhesives are perfect for quick repair jobs
  • A flexible, tough and durable, vibration resistant adhesive
  • High peel and shear strengths
  • Bonds to polystyrene, ABS nylon, metal, wood, masonry and rubber
More About this Item
Epoxies and urethanes come in handy 3.5-gram, two-component packets with mixing sticks. Choose from individual packets or two kit options providing an assortment of epoxies and urethanes.