Hardman Double Bubble® Epoxy Pouches, Machineable Fast Setting, 3.5g; 10/Pk
Representative image only.
Hardman Mfr # 4002-BG10 – Item # EW-08778-10
Complete selection of epoxies and urethane for all your maintenance needs
  • Single-use adhesives are perfect for quick repair jobs
  • A machineable tooling and body patch that adheres to metal, wood and hard plastics
  • Can be machined, sanded, drilled and tapped
PRODUCT OPTIONS
$37.50USD / Pkg of 10
BUY MORE & SAVE
  • 1-9$37.50USD / Pkg of 10
  • 10+$33.75USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Curing time (days)14
  • Curing time (hrs)336
  • Set time (mins)25
  • Set time (hrs)0.42
  • Min operating temperature (° F)-20
  • Max operating temperature (° F)300
  • Number of Pieces per Set10
  • DescriptionEpoxy Pouches, Machineable Fast Setting, 3.5g; 10/Pk
Includes
10 pouches and mixing sticks.
Key Features
  • Single-use adhesives are perfect for quick repair jobs
  • A machineable tooling and body patch that adheres to metal, wood and hard plastics
  • Can be machined, sanded, drilled and tapped
More About this Item
Epoxies and urethanes come in handy 3.5-gram, two-component packets with mixing sticks. Choose from individual packets or two kit options providing an assortment of epoxies and urethanes.