Hardman Double Bubble® Epoxy Pouches, Extra Fast Setting, 3.5g; 10/Pk
Representative image only.
Hardman Mfr # 4001-BG10 – Item # EW-08778-02
Complete selection of epoxies and urethane for all your maintenance needs
  • Single-use adhesives are perfect for quick repair jobs
  • Excellent adhesive for bonding wood, glass, metal, stone, concrete, ceramics and leather
PRODUCT OPTIONS
$44.00USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Curing time (days)1
  • Curing time (mins)1440
  • Curing time (hrs)24
  • Set time (mins)5
  • Min operating temperature (° F)-20
  • Max operating temperature (° F)180
  • Number of Pieces per Set10
  • DescriptionEpoxy Pouches, Extra Fast Setting, 3.5g; 10/Pk
Includes
10 pouches and mixing sticks.
Key Features
  • Single-use adhesives are perfect for quick repair jobs
  • Excellent adhesive for bonding wood, glass, metal, stone, concrete, ceramics and leather
More About this Item
Epoxies and urethanes come in handy 3.5-gram, two-component packets with mixing sticks. Choose from individual packets or two kit options providing an assortment of epoxies and urethanes.