Hand-Held Tubing Cutter, PTFE Coated Steel, Max 1-1/4" OD Tubing
Representative image only.
Item # EW-06438-90
Get clean, straight cuts everytime
  • Cuts through tubing up to 1 1/4" OD
$57.90USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Minimum ID (in)0.404
  • Minimum ID (mm)10.3
  • MaterialPTFE-coated Steel
  • Max Tubing OD (in)1 1/4
  • Max Tubing OD (mm)31.75
  • DescriptionHand-Held Tubing Cutter, PTFE coated steel, max 1-1/4" OD tubing
Key Features
  • Cuts through tubing up to 1 1/4" OD
More About this Item
Steel blade coated with PTFE® resin produces clean, straight cuts through tubing up to 1-1/4" OD (cuts polyethylene tubing up to 1-1/2" OD). Optional carrying pouch and replacement blade available.
Steel blade coated with PTFE® resin produces clean, straight cuts through tubing up to 1-1/4" OD (cuts polyethylene tubing up to 1-1/2" OD). Optional carrying pouch available.