Hach PH STRIPS PH Paper Test Strips, 0 to 14 PH Range; 100 Strips/Pk
Representative image only.
Hach Mfr # 2601300 – Item # EW-05546-42
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$29.39USD / Each
Often Bought with…
SPECS & DESCRIPTION
Specifications & Description
  • TestpH
  • Measurement Points0 to 14 pH in 1 pH unit increments
  • Reagent PlatformStrips
  • Tests Per Kit100
  • DescriptionPH Paper Test Strips, 0 to 14 PH Range; 100 Strips/Pk
Includes
test strips and color key for reading results.
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Test full range pH ( 0 - 14). 100 test strips included.