Hach 2751340 Chloride Test Strips High Range , 40
Representative image only.
Hach Mfr # 2751340 – Item # EW-05546-18
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$81.80USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestChloride, high range
  • Measurement Points300 to 6000 ppm
  • Reagent PlatformStrips
  • Tests Per Kit40
  • DescriptionChloride Test Strips High Range , 40
ISO9001 Certified Supplier
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Tests chloride as Cl- from 300 to 6000 mg/L. 40 test strips included.