Hach 2745650 PH MidRange (4 - 9) Test Strips 50 (Graduations 4, 5, 6, 7, 8, 9)
Representative image only.
Hach Mfr # 2745650 – Item # EW-05546-40
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$20.25USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestpH
  • Measurement Points4 to 9 pH in 1 pH unit increments
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionPH MidRange (4 - 9) Test Strips 50 (Graduations 4, 5, 6, 7, 8, 9)
ISO9001 Certified Supplier
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Test pH in midrange of 4 to 9 pH units. 50 test strips included.