Hach 2745250 Total Hardness Test Strips, 0 to 425 mg/L; 50 Strips/Pk
Representative image only.
Hach Mfr # 2745250 – Item # EW-05546-30
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$20.40USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestHardness, calcium carbonate
  • Measurement Points0, 25, 50, 120, 250, 425 ppm
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionTotal Hardness Test Strips, 0 to 425 mg/L; 50 Strips/Pk
ISO9001 Certified Supplier
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Test hardness as CaC)3 in the range of 0 to 425 mg/L ( 0 to 25 gpg). 50 individually wrapped test strips included.