Hach 2745050 Free and Total Chlorine Low Level Test Strips, 50
Representative image only.
Hach Mfr # 2745050 – Item # EW-05546-22
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$29.90USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestChlorine, free and total; low range
  • Measurement Points0, 0.5, 1, 2, 4, 10 ppm
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionFree and Total Chlorine Low Level Test Strips, 50
ISO9001 Certified Supplier
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Tests free and total chlorine as Cl2 from 0 to 10 mg/L. 50 individually wrapped test strips included.