Hach 2744850 Total Alkalinity Test Strips, 0 to 240 mg/L; 50 Strips/Pk
Representative image only.
Hach Mfr # 2744850 – Item # EW-05546-04
Reduce risk by using as a pre-test validation tool
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
PRODUCT OPTIONS
$20.65USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestAlkalinity
  • Measurement Points0, 40, 80, 120, 240 ppm
  • Reagent PlatformStrips
  • Tests Per Kit50
  • DescriptionTotal Alkalinity Test Strips, 0 to 240 mg/L; 50 Strips/Pk
ISO9001 Certified Supplier
Key Features
  • Simplified testing with results in less than one minute
  • Test strips eliminate chemical handling and cleanup
  • Vial label contains the color key for easy readings in lab or field
More About this Item
Measures total alkainity from 0 to 240 mg/L.
50 test strips included.