Hach 2712500 Speccheck Standards for Fluoride (0 to 2.00 mg/L)
Representative image only.
Hach Mfr # 2712500 – Item # EW-99574-95
Ideal for Pocket Colorimeter II calibration
  • Four vials of known concentration standards
  • 1 year warranty
PRODUCT OPTIONS
$278.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Test ParametersFluoride
  • DescriptionSpeccheck Standards for Fluoride (0 to 2.00 mg/L)
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Four vials of known concentration standards
  • 1 year warranty
More About this Item
Spec Gel Secondary Standard kit--four sealed vials of known concentration from 0 to 2.0 mg/L for Fluoride.