Hach 2291700 Hydrogen Peroxide Drop Test Kit , 10 ppm Max, 100 Tests/Kit
Representative image only.
Hach Mfr # 2291700 – Item # EW-05545-36
Low cost and simple drop count titration method
  • Kit contains all required reagents and apparatus
  • 1 year warranty
PRODUCT OPTIONS
$137.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestHydrogen peroxide
  • Measurement Points0.2 to 2 ppm; 1 to 10 ppm
  • Reagent PlatformThiosulfate
  • Tests Per Kit100
  • DescriptionHydrogen Peroxide Drop Test Kit , 10 ppm Max, 100 Tests/Kit
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations. 100 tests per pack.
Rugged carrying case for analysis in the field