Hach 2183100 formaldehyde Drop Test Kit , 10% Max, 100 Tests/Kit
Representative image only.
Hach Mfr # 2183100 – Item # EW-05545-26
Low cost and simple drop count titration method
  • Kit contains all required reagents and apparatus
  • 1 year warranty
PRODUCT OPTIONS
$159.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestFormaldehyde
  • Measurement Points0.05 to1%; 0.5 to 10%
  • Reagent PlatformThymolphthalein
  • Tests Per Kit100
  • Descriptionformaldehyde Drop Test Kit , 10% Max, 100 Tests/Kit
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations. 100 tests per pack.
Rugged carrying case for analysis in the field