Hach 146900 Dissolved Oxygen Drop Test Kit ( 0.2 to 4; 1 to 20 ppm)
Representative image only.
Hach Mfr # 146900 – Item # EW-05545-24
Low cost and simple drop count titration method
  • Kit contains all required reagents and apparatus
  • 1 year warranty
PRODUCT OPTIONS
$143.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestDissolved oxygen
  • Measurement Points0.2 to 4 ppm; 1 to 20 ppm
  • Reagent PlatformModified Winkler
  • Tests Per Kit100
  • DescriptionDissolved Oxygen Drop Test Kit ( 0.2 to 4; 1 to 20 ppm)
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations. 100 tests per pack.
Rugged carrying case for analysis in the field