Hach 1455400 Silica, High Range Color Disc Teat Kit
Representative image only.
Hach Mfr # 1455400 – Item # EW-05545-51
Color wheel method provides accurate visual comparisons
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
PRODUCT OPTIONS
$215.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestSilica, high range
  • Measurement Points 0 to 30 ppm; 25 to 800 ppm
  • Tests Per Kit100
  • DescriptionSilica, High Range Color Disc Teat Kit
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Quick and simple water testing results in an easy-to-carry kit
  • 1 year warranty
More About this Item
Color disc test kits have a color wheel to make visual test matches to estimate concentration in water.