Hach 144001 Chloride Drop Test Kit , Low Range ( 5 to 100; 20 to 400 ppm)
Representative image only.
Hach Mfr # 144001 – Item # EW-05545-14
Low cost and simple drop count titration method
  • Kit contains all required reagents and apparatus
  • 1 year warranty
PRODUCT OPTIONS
$91.90USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestChloride, low range
  • Measurement Points5 to 100 ppm; 20 to 400 ppm
  • Reagent PlatformSilver Nitrate
  • Tests Per Kit100
  • DescriptionChloride Drop Test Kit , Low Range ( 5 to 100; 20 to 400 ppm)
  • Warranty1 year
1 Year WarrantyISO9001 Certified Supplier
Key Features
  • Kit contains all required reagents and apparatus
  • 1 year warranty
More About this Item
Drop count titration is a simple method to estimate concentrations. 100 tests per pack.
Rugged carrying case for analysis in the field