H-B Instrument B60900-2800 DURAC Probeless Electronic Indoor/Outdoor Thermometer; -40/50C (-40/122F)
Representative image only.
H-B Instrument Mfr # 609002800 – Item # EW-08081-12
Simultaneously displays current, maximum, and minimum temperatures
  • Switch between indoor/outdoor temperature readings
  • Super-large LCD
  • 1 year warranty
$90.80USD / Each
$191.50 - $264.00USD / Each
for an additional
$191.50USD / Each
for an additional
$264.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeDigital Indicator
  • Min Temperature (° F)-40
  • Max Temperature (° F)122
  • Min Temperature (° C)-40
  • Max Temperature (° C)50
  • Resolution0.1°
  • DescriptionInstrument Digital Min/Max Indoor /Outdoor Thermometer 08081-12
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Switch between indoor/outdoor temperature readings
  • Super-large LCD
  • 1 year warranty
More About this Item
The meter simultaneously displays current, maximum, and minimum temperatures with the ability to switch between indoor and outdoor temperature modes. Readings are shown on an extra large LCD that is easily viewed from several feet away, with the current temperature displayed by a bar scale. The unit features a Min/Max function that offers the highest and lowest readings from when the reset button was last pressed. Other features include a remote probe for outdoor readings, high/low alarm, low battery indicator, and ºF/°C selection.