Great Plains Industries Flowmeter/Totalizer, 5 to 50 GPM; 1" male spigot connection
Representative image only.
Great Plains Industries Mfr # TM10SQ9GMA – Item # EW-05611-12
Local display includes rate of flow, batch, and cumulative totals
  • Heavy-duty schedule 80 PVC construction
PRODUCT OPTIONS
$425.40USD / Each
$332.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±3.0
  • Min Flow Rate (GPM)5
  • Max Flow Rate (GPM)50
  • Wetted MaterialsPVC, ceramic, tungsten cabide, PVDF, 316 SS
  • Min Operating Temperature (° F)32
  • Min Operating Temperature (° C)0
  • Max Operating Temperature (° F)140
  • Max Operating Temperature (° C)60
  • Process Connection1" male spigot
  • Fluid Typewater and thin fluids
  • BatteryTwo lithium batteries
  • Battery Life9000 hours
  • Display Type6-digit LCD, 0.5"H
  • DescriptionFlowmeter/Totalizer, 5 to 50 GPM; 1" male spigot connection
Key Features
  • Heavy-duty schedule 80 PVC construction
More About this Item
These durable meters offer accuracy at a reasonable price for measuring water and are available in flow ranges from 1 to 200 GPM and 1/2'' to 2'' inch fitting sizes. Meters indicate totalization and rate of flow, and read up to 999,999 gallons or liters — switch between gallon and liter and the electronics will indicate accurately in both units of measure. Models 05611-10 to -19 offer digital indication powered by two replaceable lithium batteries. Models 05611-50 to -59 have no display and have a square wave pulse output. Flowmeters are available with NPT or male spigot fittings, and work perfect for existing PVC plumbed systems. Wetted parts include the PVC housing, tungsten carbide shaft, PVDF rotor and supports and ceramic bearings. Flowmeters are field calibrateable.