Great Plains Industries Flowmeter Remote Mountng Kit with 10 ft (3 m) Cable
Representative image only.
Great Plains Industries Mfr # 113275-1 – Item # EW-05609-93
Make the most of your Great Plains Industries flowmeters with these replacement parts and other accessories
  • Optimize performance of your flowmeter with these accessories
PRODUCT OPTIONS
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionFlowmeter Remote Mountng Kit with 10 ft (3 m) Cable
Key Features
  • Optimize performance of your flowmeter with these accessories
More About this Item
These accessories and replacement parts are suitable for use with your Great Plains Industries flowmeters.
This remote kit modifies all GPI and Cole-Parmer Heavy-Duty Turbine Flowmeters/Totalizers for when you need remote indication. The kit provides versatility—mount the LCD readout up to 100 ft (30 m) from the flowmeter! The kit consists of a sensor module and 10-ft of cable.