GF Signet P525-2 Metalex Paddle-Wheel Flow Sensors, for use with mini-tap fitting; 2-1/2"
Representative image only.
GF Signet Mfr # P525-2 – Item # EW-05618-64
Stainless steel body withstands high pressures and temperatures
  • Use in agressive liquid flow applications
PRODUCT OPTIONS
$743.00USD / Each
$310.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Repeatability±0.5% full-scale
  • Length (")2 1/2
  • Max Operating Temperature (° F)300
  • Max Operating Temperature (° C)149
  • DescriptionMetalex Paddle-Wheel Flow Sensors, for use with mini-tap fitting; 2-1/2"
Includes
25-foot signal cable
Key Features
  • Use in agressive liquid flow applications
More About this Item
Use these rugged sensors to monitor the flow of liquid ammonia, turbine steam condensate, and more—wherever strength and chemical resistance are required.
Open-core rotor is cavitation free at flow rates from 1.5 to 20 ft/second. Easy maintenance—sensors can be replaced in minutes.
METALEX flow sensors are compatible with all of our battery- and line-powered flowmeters, accumulators, and controllers. Sensors must be used with the fitting type specified in the table below—installation fittings sold separately.
METALEX sensors feature CD4MCU rotor; Fluoroloy B rotor bearings; and 316 stainless steel body, rotor shaft, and shaft retainers.