GF Signet 3-8050-2 Junction Box With Easycal ( )
Representative image only.
GF Signet Mfr # 3-8050-2 – Item # EW-68476-67
Junction boxes available for convenient wiring
  • 2 year warranty
PRODUCT OPTIONS
$196.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionJunction Box With Easycal ( )
  • Warranty2 Years
Two year warranty
Key Features
  • 2 year warranty
More About this Item
Junction boxes simplify wiring and enable cable extension of pH or ORP sensors.