GF Signet 3-2823-1 Conductivity/Resistivity Cells; range, 200 - 400,000 mS; fitting material, 316 SS
Representative image only.
GF Signet Mfr # 3-2823-1 – Item # EW-19905-53
Designed to provide a versatile installation and accurate sensing across a broad range
  • Wetted Materials: 316 SST or Titanium
  • Thread is reversible for submersion mounting
  • 1 year warranty
$1,234.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Cell ConstantK = 20
  • Body Material316 SS
  • DescriptionConductivity/Resistivity Cells; range, 200 - 400,000 mS; fitting material, 316 SS
  • Warranty1 year
1 Year Warranty
Key Features
  • Wetted Materials: 316 SST or Titanium
  • Thread is reversible for submersion mounting
  • 1 year warranty
More About this Item
Compatible Instruments: Signet Instrument Types: 8850, 8860 8900 or the 9900 with the 2850 Sensor Electronics