General Tools PIN4E Replacement Pins for MM1E and MMH800 (2 Pair)
Representative image only.
General Tools Mfr # PIND4E – Item # EW-37800-19
A must-have tool for woodworking, building construction, and home inspection
  • Check for moisture in carpets and subflooring
  • Great for identifying leaks in roofs, floors, and walls
  • 10-LED dual-scale readout with separate High/Low indicators
  • 1 year warranty
PRODUCT OPTIONS
$11.39USD / Each
$13.40USD / Each
Include Meter Today!
SPECS & DESCRIPTION
Specifications & Description
  • Measurement TypeResistive
  • DescriptionReplacement Pins for MM1E and MMH800 (2 Pair)
  • Warranty1 year
1 Year Warranty
Key Features
  • Check for moisture in carpets and subflooring
  • Great for identifying leaks in roofs, floors, and walls
  • 10-LED dual-scale readout with separate High/Low indicators
  • 1 year warranty
More About this Item
Use this low-cost meter with pins for a surface moisture measurement in building materials such as wood, plaster, drywall, concrete, roofing, and insulation. A protective cap doubles as a calibration checker. Pins provide 3/8” (1 cm) penetration for testing soft materials such as soil, paper, or powders. Simply replace the pins when they wear out. Unit features full- and low-battery indicators.