Fuego 8.004.000 Scs Pro -- with Infrared and Rechargeable Battery

Representative image only.
Fuego Mfr # 8 204 000 – Item # EW-36302-06
Simple and safe operation for all flame related applications in the laboratory
  • Ideal for use in all industrial, laboratory, and cleanroom settings
  • Modern low-profile body design reduces air flow disturbances and is very easy to clean
  • Robust, all stainless steel construction and fireproof controls
  • 2 year warranty

PRODUCT OPTIONS

$1,241.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Gas TypeNatural Gas, Propane/Butane
  • Inlet Tube OD (in)0.25
  • Width (in)5 1/2
  • Length (in)4
  • Height (in)1 15/16
  • Power (VAC)120 / 240
  • DescriptionScs Pro -- with Infrared and Rechargeable Battery
  • Warranty2 Years
Two year warrantyUL Listed to Canadian safety standards.Conformite Europeenne

Key Features

  • Ideal for use in all industrial, laboratory, and cleanroom settings
  • Modern low-profile body design reduces air flow disturbances and is very easy to clean
  • Robust, all stainless steel construction and fireproof controls
  • 2 year warranty

More About this Item

With an ergonomic design and extensive standard features, these laboratory gas burners withstand extreme lab conditions while incorporating innovative safety technology. The Safety Control System (SCS) constantly analyzes potential hazards, and, if necessary, initiates safety measures, such as an interruption of the gas supply. An adjustable automatic cut-off system inhibits unintentional ignition. To protect you from burns after prolonged use, the residual heat display will signal that the burner head is still hot. Burners also feature continuous burner head control which identifies burner head clogging and ensures correct assembly of burner head.

Fuego SCS Pro Burner offers all the features of the above models plus adds maximum flexibility through the state-of-the-art rechargeable battery technology. Unit includes an integrated fast charge function and two rechargeable batteries ( NiMH 2.4V, Mignon (AA)) for up to 9 hours of continuous use.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat