Fowler 54-004-854 IP54 Electronic 6" Caliper and 1" Micrometer Set
Representative image only.
Fowler Mfr # 540048540 BOM – Item # EW-97604-40
  • 1 year warranty
$600.50USD / Each
$43.80 - $116.50USD / Each
for an additional
$43.80USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionIP54 Electronic 6\" Caliper and 1\" Micrometer Set
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Toolmaker Universal Measuring Set
  • 6" dial caliper-.001" graduations
  • 0-1" micrometer - .0001" graduations
  • 5" flexible rule with 32nds, 64ths, 10ths and 100ths graduatiosn
Shockproof Measuring Set
  • 0-6" shockproof caliper, .100" range per revolution with .001" reading dial face
  • 0-1" micrometer with .0001" graduatiosn and ratchet thimble
  • 5R steel rule
Electronic Caliper and Micrometer Measuring Set
  • 6" electronic caliper - .0005"/.01mm resolution with ceramic chip
  • 0-1"/25mm electronic micrometer - .00005"/.001mm resolution
Water Resistant Electronic Measuring Set
  • Both caliper and micrometer are protected to IP54 standards
  • 0-6" / 150mm Ultra-Cal IV Caliper
  • 0-1" micrometer
  • 4R steel rule