Fluke I1010 Current Probe AC/DC Clamp-On
Representative image only.
Fluke Mfr # I1010 – Item # EW-20003-40
Sold only in the United States and Canada
  • Included with kit:
  • i1010 1-600A AC rms, 1-1000A DC Current Clamp
  • Zippered vinyl carry case with moveable divider
  • 1 year warranty
PRODUCT OPTIONS
$481.49USD / Each
$534.99USD / Each
$112.25 - $185.00USD / Each
for an additional
$112.25USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • AC Max (amps)600
  • Safety RatingCAT III 600V
  • DescriptionCurrent Probe AC/DC Clamp-On
  • Warranty1 Year
1 Year Warranty
Key Features
  • Included with kit:
  • i1010 1-600A AC rms, 1-1000A DC Current Clamp
  • Zippered vinyl carry case with moveable divider
  • 1 year warranty
More About this Item
AC Current Clamps and Probes for use with DMMS, ScopeMeters and Power Quality Analyzers
Special value kit for electrical professional who is looking to add on DC and AC current measurement capabilities to their DMM. Includes a roomy carry case to hold and protect your meter and current clamp, plus extra storage for your test leads, probes and manuals.