Fluke FLK-V3001 FC Wireless DC Voltage Module

Representative image only.
Fluke Mfr # FLK-V3001 FC – Item # EW-20047-81
See, save, and share data wirelessly — saving time and increasing productivity
  • Increase productivity by communicating wirelessly between your phone, PC, and Fluke test tools
  • Download the Fluke Connect™ app for use with iOS (4s and later) and Android™ to connect with over twenty Fluke test tools (Download iOS app at the App Store, and the Android app from Google play)
  • Logging function on modules allow you to isolate intermittent events or record signal fluctuations remotely
  • Share data with associates in seconds, allowing for immediate interpretation and action
  • Increase efficiency by simultaneously viewing multiple measurements on one screen
  • Take measurements from safe locations reducing potential hazards
  • 3 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • DC Min (Volts)0.0001
  • DC Max (Volts)1000
  • DC Voltage Accuracy0.09% ±3 digits
  • Width (in)2.33
  • Height (in)6.33
  • Depth (in)1.5
  • Width (cm)6.6
  • Height (cm)16
  • Depth (cm)3.8
  • DescriptionWireless DC Voltage Module
  • Warranty3 Years
3 Year WarrantyConformite EuropeenneCanadian Standards Association

Key Features

  • Increase productivity by communicating wirelessly between your phone, PC, and Fluke test tools
  • Download the Fluke Connect™ app for use with iOS (4s and later) and Android™ to connect with over twenty Fluke test tools (Download iOS app at the App Store, and the Android app from Google play)
  • Logging function on modules allow you to isolate intermittent events or record signal fluctuations remotely
  • Share data with associates in seconds, allowing for immediate interpretation and action
  • Increase efficiency by simultaneously viewing multiple measurements on one screen
  • Take measurements from safe locations reducing potential hazards
  • 3 year warranty

More About this Item

The Fluke V3001 FC measures DC voltage from 0.1 mV to 1000 V and can be used as a standalone meter or as part of the Fluke Connect wireless system. The logging function records and saves up to 65,000 readings for added flexibility.

The Fluke V3001 FC is a fully functional DC voltage meter that wirelessly relays DC voltage measurements to other Fluke Connect enabled master units listed below:
Fluke 3000 FC DMM
Fluke Ti200/300/400 Infrared Cameras
To your PC via optional PC3000 FC adapter

What’s included: TL222 test leads, AC220 alligator clips, magnetic hanging strap, and two AA batteries.

Available for sale in the United States and Canada only

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat