Fluke Calibration 9118A-3118A-1 Isothermal Block for T/C Calibration Furnace

Representative image only.
Fluke Calibration Mfr # 9118A-3118A-1 – Item # EW-90914-06

$2,219.40USD / Each
$2,466.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Temperature Accuracy±9°F (±5°C)
  • Stability±0.1°C (±0.2°F)
  • DescriptionIsothermal Block for T/C Calibration Furnace

More About this Item

Various parts and accessories to change capacity, testing type, accuracy, or portability for your Fluke Calibration calibrator.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat