Fluke Calibration 3376859 Thermistor Probe; 1/8" Dia x 6" Linfo-Con 1523/24
Representative image only.
Fluke Calibration Mfr # 5610-6-P – Item # EW-90451-06
  • 1 year warranty
$1,336.00USD / Each
Time to recalibrate?
for an additional
$520.00USD / Each
for an additional
$593.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Probe TypeThermistor
  • Min Temperature (° F)32
  • Max Temperature (° F)212
  • Min Temperature (° C)0
  • Max Temperature (° C)100
  • Temperature Accuracy±0.018°F (±0.01°C)
  • Connector TypeInfo-Con
  • Sheath Diameter (in)0.125
  • DescriptionThermistor Probe; 1/8\" Dia x 6\" Linfo-Con 1523/24
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
5610 Series - Secondary thermistor probes: The small drift and low calibration uncertainty of only ±0.01°C makes this rugged stainless steel thermistor a wise choice when temperatures of interest are between 0 and 100°C. These probes are ideal for measuring temperature in air, liquid or soil.
Available for sale in the United States and Canada only.