Fluke 87V High Accuracy True-RMS Digital Multimeter

Representative image only.
Fluke Mfr # FLUKE-87-5 – Item # EW-26087-02
Diagnostic functions provide more troubleshooting power
  • Unique function for accurate voltage and frequency measurements on adjustable speed motor drives and other electrically noisy equipment
  • True RMS AC voltage and current for accurate measurements on nonlinear signals
  • Built-in thermometer conveniently allows you to take temperature readings without having to carry a separate instrument
  • Measure up to 1000 VAC and DC
  • Measure up to 10 A (20 A for 30 seconds)

$413.99USD / Each
$459.99USD / Each

$94.00USD / Each

for an additional
$94.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Add a hard shell carrying case to protect your meter.

SPECS & DESCRIPTION

Specifications & Description

  • DC Max (Volts)1000
  • DC Voltage Accuracy±0.05% 1
  • DC Max (amps)10
  • DC Current Accuracy±0.2% 2
  • Current (AC/DC)10 A
  • AC Max (Volts)1000
  • Ac Voltage Accuracy±0.7% 2 True RMS
  • AC Max (amps)10
  • AC Current Accuracy±1.0% 2 True RMS
  • Resistance Min (Ω)600
  • Resistance Max (MΩ)50
  • Capacitance Accuracy±1.0% 2
  • Frequency Max (KHz)199.99
  • Frequency Accuracy±0.005% 1
  • Resistivity Accuracy±0.2% 1
  • Temperature Accuracy1.0%
  • Count20000
  • Digits4.5
  • Conductance50
  • Safety RatingCAT III 1000V, CAT III 600V
  • DescriptionHigh Accuracy True-RMS Digital Multimeter

Includes

TL75 test leads, AC72 alligator clips, temperature probe, CD-ROM (user's manual and technical notes), operator's guide, holster, and 9 V battery (installed).

Key Features

  • Unique function for accurate voltage and frequency measurements on adjustable speed motor drives and other electrically noisy equipment
  • True RMS AC voltage and current for accurate measurements on nonlinear signals
  • Built-in thermometer conveniently allows you to take temperature readings without having to carry a separate instrument
  • Measure up to 1000 VAC and DC
  • Measure up to 10 A (20 A for 30 seconds)
  • 10,000 µF capacitance range
  • Frequency to 200 kHz and % duty cycle
  • Peak capture to record transients as fast as 250 µs
  • Large display digits and 2-level bright white backlight for increased visibility
  • Withstands hazardous 8000 volt spikes caused by load switching and faults on industrial circuits and complies with second edition ANSI and IEC electrical safety standards
  • Improved sleep mode for long battery life
  • Relative mode to remove test lead resistance from low ohms measurements
  • Auto and manual ranging for maximum flexibility
  • Optional magnetic hanger (TPAK) for easy setup and viewing while freeing your hands for other tasks

More About this Item

Available for sale in the United States and Canada only

The Fluke 87V has improved measurement functions, troubleshooting features, resolution and accuracy to solve more problems on motor drives, in plant automation, power distribution, and electromechanical equipment. The 87V provides more problem-solving power, safety, convenience and impact protection.

The 87V is CAT III 1000 V and CAT IV 600 V rated and can withstand 8000 volt spikes caused by load switching on industrial circuits. Meter is auto and manual ranging for maximum flexibility during testing. Measurements display numerically and graphically with analog bar graph on the two-level, backlit LCD. Min/Max and average readings are recorded to capture variations. Meter features True-RMS measurements with the unique function for accurate voltage and frequency measurements on adjustable speed motor drives and other electrically noisy equipment. Unit also features a built-in temperature probe for temperature measurement capability.

ACCESSORIES

Accessories

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat